Anti-PAG1 Mouse Monoclonal Antibody [clone: 504004]

Supplier: R & D Systems
MAB5285
102885-938EA 566.89 USD
102885-938
Anti-PAG1 Mouse Monoclonal Antibody [clone: 504004]
Antibodies
Simple Western

Type: Primary
Antigen: PAG1
Clonality: Monoclonal
Clone: 504004
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR