Anti-GRM8 Mouse Monoclonal Antibody [clone: 476410]

Supplier: R & D Systems
MAB5277
220021-131EA 577.02 USD
220021-131
Anti-GRM8 Mouse Monoclonal Antibody [clone: 476410]
Antibodies
Type: Primary
Antigen Symbol: GRM8
Antigen Name: glutamate receptor, metabotropic 8
Antigen Synonyms: GLUR8, GPRC1H, MGLUR8, mGlu8
Clonality: Monoclonal
Clone: 476410
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR