Anti-TPH1 Mouse Monoclonal Antibody [clone: 508417]

Supplier: R & D Systems
MAB5276
220021-130EA 577.02 USD
220021-130
Anti-TPH1 Mouse Monoclonal Antibody [clone: 508417]
Antibodies
Type: Primary
Antigen: TPH1
Clonality: Monoclonal
Clone: 508417
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR