Anti-CLDN8 Mouse Monoclonal Antibody [clone: 474210]

Supplier: R & D Systems
MAB5275
220021-129EA 594.37 USD
220021-129
Anti-CLDN8 Mouse Monoclonal Antibody [clone: 474210]
Antibodies
Type: Primary
Antigen: CLDN8
Clonality: Monoclonal
Clone: 474210
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR