Anti-MET Rat Monoclonal Antibody [clone: 118624]

Supplier: R & D Systems
MAB527
220021-126EA 763.57 USD
220021-126
Anti-MET Rat Monoclonal Antibody [clone: 118624]
Antibodies
Type: Primary
Antigen: MET
Clonality: Monoclonal
Clone: 118624
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR