Anti-NOTCH1 Mouse Monoclonal Antibody [clone: N1A]

Supplier: R & D Systems
MAB5267
220021-125EA 577.02 USD
220021-125
Anti-NOTCH1 Mouse Monoclonal Antibody [clone: N1A]
Antibodies
Type: Primary
Antigen: NOTCH1
Clonality: Monoclonal
Clone: N1A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR