Anti-IL34 Mouse Monoclonal Antibody [clone: 578410]

Supplier: R & D Systems
MAB52651
102885-936EA 772.25 USD
102885-936
Anti-IL34 Mouse Monoclonal Antibody [clone: 578410]
Antibodies
Type: Primary
Antigen: IL34
Clonality: Monoclonal
Clone: 578410
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR