Anti-IL34 Mouse Monoclonal Antibody [clone: 578416]

Supplier: R & D Systems
MAB5265
220021-124EA 594.37 USD
220021-124
Anti-IL34 Mouse Monoclonal Antibody [clone: 578416]
Antibodies
Type: Primary
Antigen: IL34
Clonality: Monoclonal
Clone: 578416
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR