Anti-HHV8-ORF74 Mouse Monoclonal Antibody [clone: 462510]

Supplier: R & D Systems
MAB5264
220021-123EA 577.02 USD
220021-123
Anti-HHV8-ORF74 Mouse Monoclonal Antibody [clone: 462510]
Antibodies
Type: Primary
Antigen: HHV8-ORF74
Clonality: Monoclonal
Clone: 462510
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Viral
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR