Anti-FASLG Rat Monoclonal Antibody [clone: 101626]

Supplier: R & D Systems
MAB5262
220021-122EA 763.57 USD
220021-122
Anti-FASLG Rat Monoclonal Antibody [clone: 101626]
Antibodies
Type: Primary
Antigen Symbol: FASL
Antigen Name: Fas ligand (TNF superfamily, member 6)
Antigen Synonyms: APT1LG1, CD178, CD95-L, CD95L, Fas-L, Faslg, Tnfsf6, gld
Clonality: Monoclonal
Clone: 101626
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR