Anti-IFNLR1 Mouse Monoclonal Antibody [clone: 601106]

Supplier: R & D Systems
MAB5260
102885-934EA 616.06 USD
102885-934
Anti-IFNLR1 Mouse Monoclonal Antibody [clone: 601106]
Antibodies
Type: Primary
Antigen: IFNLR1
Clonality: Monoclonal
Clone: 601106
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR