Anti-CXCL2 Mouse Monoclonal Antibody [clone: 123802]

Supplier: R & D Systems
MAB525
220021-118EA 763.57 USD
220021-118
Anti-CXCL2 Mouse Monoclonal Antibody [clone: 123802]
Antibodies
Type: Primary
Antigen: CXCL2
Clonality: Monoclonal
Clone: 123802
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR