Anti-CD24 Mouse Monoclonal Antibody [clone: 528807]

Supplier: R & D Systems
MAB5248
220021-117EA 594.37 USD
220021-117
Anti-CD24 Mouse Monoclonal Antibody [clone: 528807]
Antibodies
Type: Primary
Antigen: CD24
Clonality: Monoclonal
Clone: 528807
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR