Anti-HCRTR2 Mouse Monoclonal Antibody [clone: 456738]

Supplier: R & D Systems
MAB5246
220021-115EA 577.02 USD
220021-115
Anti-HCRTR2 Mouse Monoclonal Antibody [clone: 456738]
Antibodies
Type: Primary
Antigen: HCRTR2
Clonality: Monoclonal
Clone: 456738
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR