Anti-TYROBP Mouse Monoclonal Antibody [clone: 406288]

Supplier: R & D Systems
MAB5240
220021-113EA 633.42 USD
220021-113
Anti-TYROBP Mouse Monoclonal Antibody [clone: 406288]
Antibodies
Type: Primary
Antigen: TYROBP
Clonality: Monoclonal
Clone: 406288
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR