Anti-TNFRSF18 Rat Monoclonal Antibody [clone: 108626]

Supplier: R & D Systems
MAB524
220021-112EA 772.25 USD
220021-112
Anti-TNFRSF18 Rat Monoclonal Antibody [clone: 108626]
Antibodies
Type: Primary
Antigen: TNFRSF18
Clonality: Monoclonal
Clone: 108626
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR