Anti-GPRC5A Mouse Monoclonal Antibody [clone: 481906]

Supplier: R & D Systems
MAB5239
220021-111EA 623.29 USD
220021-111
Anti-GPRC5A Mouse Monoclonal Antibody [clone: 481906]
Antibodies
Type: Primary
Antigen Symbol: GPRC5A
Antigen Name: G protein-coupled receptor, class C, group 5, member A
Antigen Synonyms: GPCR5A, PEIG-1, RAI3, RAIG1, TIG1
Clonality: Monoclonal
Clone: 481906
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR