Anti-SEMA4D Rat Monoclonal Antibody [clone: 547412]

Supplier: R & D Systems
MAB52351
102885-932EA 561.11 USD
102885-932
Anti-SEMA4D Rat Monoclonal Antibody [clone: 547412]
Antibodies
Type: Primary
Antigen: SEMA4D
Clonality: Monoclonal
Clone: 547412
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR