Anti-SEMA4D Rat Monoclonal Antibody [clone: 547406]

Supplier: R & D Systems
MAB5235
220021-110EA 594.37 USD
220021-110
Anti-SEMA4D Rat Monoclonal Antibody [clone: 547406]
Antibodies
Type: Primary
Antigen: SEMA4D
Clonality: Monoclonal
Clone: 547406
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR