Anti-CRTAC1 Mouse Monoclonal Antibody [clone: 755339]

Supplier: R & D Systems
MAB52341
102885-930EA 566.89 USD
102885-930
Anti-CRTAC1 Mouse Monoclonal Antibody [clone: 755339]
Antibodies
Type: Primary
Antigen: CRTAC1
Clonality: Monoclonal
Clone: 755339
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR