Anti-IL18 Mouse Monoclonal Antibody [clone: 69604]

Supplier: R & D Systems
MAB521
220021-107EA 772.25 USD
220021-107
Anti-IL18 Mouse Monoclonal Antibody [clone: 69604]
Antibodies
Type: Primary
Antigen: IL18
Clonality: Monoclonal
Clone: 69604
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR