Anti-NOTCH2 Rat Monoclonal Antibody [clone: 605724]

Supplier: R & D Systems
MAB5196
102885-926EA 577.02 USD
102885-926
Anti-NOTCH2 Rat Monoclonal Antibody [clone: 605724]
Antibodies
Type: Primary
Antigen: NOTCH2
Clonality: Monoclonal
Clone: 605724
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR