Anti-EPHA10 Mouse Monoclonal Antibody [clone: 494130]

Supplier: R & D Systems
MAB5188
102885-922EA 594.37 USD
102885-922
Anti-EPHA10 Mouse Monoclonal Antibody [clone: 494130]
Antibodies
Type: Primary
Antigen: EPHA10
Clonality: Monoclonal
Clone: 494130
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR