Anti-CDNF Rat Monoclonal Antibody [clone: 560108]

Supplier: R & D Systems
MAB5187
102885-920EA 561.11 USD
102885-920
Anti-CDNF Rat Monoclonal Antibody [clone: 560108]
Antibodies
Type: Primary
Antigen: CDNF
Clonality: Monoclonal
Clone: 560108
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR