Anti-ANXA6 Mouse Monoclonal Antibody [clone: 686921]

Supplier: R & D Systems
MAB5186
102885-918EA 577.02 USD
102885-918
Anti-ANXA6 Mouse Monoclonal Antibody [clone: 686921]
Antibodies
Type: Primary
Antigen: ANXA6
Clonality: Monoclonal
Clone: 686921
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR