Anti-CSF2 Mouse Monoclonal Antibody [clone: 83308]

Supplier: R & D Systems
MAB5181
220021-105EA 772.25 USD
220021-105
Anti-CSF2 Mouse Monoclonal Antibody [clone: 83308]
Antibodies
Type: Primary
Antigen: CSF2
Clonality: Monoclonal
Clone: 83308
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR