Anti-CSF2 Mouse Monoclonal Antibody [clone: 83329]

Supplier: R & D Systems
MAB518
220021-104EA 772.25 USD
220021-104
Anti-CSF2 Mouse Monoclonal Antibody [clone: 83329]
Antibodies
Type: Primary
Antigen: CSF2
Clonality: Monoclonal
Clone: 83329
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR