Anti-THSD1 Mouse Monoclonal Antibody [clone: 541213]

Supplier: R & D Systems
MAB5178
220021-103EA 561.11 USD
220021-103
Anti-THSD1 Mouse Monoclonal Antibody [clone: 541213]
Antibodies
Type: Primary
Antigen Symbol: THSD1
Antigen Name: thrombospondin, type I, domain containing 1
Antigen Synonyms: TMTSP, UNQ3010
Clonality: Monoclonal
Clone: 541213
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR