Anti-UMOD Rat Monoclonal Antibody [clone: 774056]

Supplier: R & D Systems
MAB5175
102885-916EA 600.15 USD
102885-916
Anti-UMOD Rat Monoclonal Antibody [clone: 774056]
Antibodies
Type: Primary
Antigen: UMOD
Clonality: Monoclonal
Clone: 774056
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR