Anti-CLEC3B Mouse Monoclonal Antibody [clone: 805035]

Supplier: R & D Systems
MAB5170
102885-914EA 763.57 USD
102885-914
Anti-CLEC3B Mouse Monoclonal Antibody [clone: 805035]
Antibodies
Blockade of Receptor-ligand Interaction, Simple Western

Type: Primary
Antigen: CLEC3B
Clonality: Monoclonal
Clone: 805035
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR