Anti-ENO2 Mouse Monoclonal Antibody [clone: 522601]

Supplier: R & D Systems
MAB5169
220021-101EA 660.89 USD
220021-101
Anti-ENO2 Mouse Monoclonal Antibody [clone: 522601]
Antibodies
Type: Primary
Antigen: ENO2
Clonality: Monoclonal
Clone: 522601
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR