Anti-S100 A11 Rat Monoclonal Antibody [clone: 493804]

Supplier: R & D Systems
MAB5167
220021-100EA 594.37 USD
220021-100
Anti-S100 A11 Rat Monoclonal Antibody [clone: 493804]
Antibodies
Type: Primary
Antigen: S100 A11
Clonality: Monoclonal
Clone: 493804
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR