Anti-CXCL3 Mouse Monoclonal Antibody [clone: 83504]

Supplier: R & D Systems
MAB516
220021-099EA 763.57 USD
220021-099
Anti-CXCL3 Mouse Monoclonal Antibody [clone: 83504]
Antibodies
Type: Primary
Antigen: CXCL3
Clonality: Monoclonal
Clone: 83504
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR