Anti-IL2RA Mouse Monoclonal Antibody [clone: 745520]

Supplier: R & D Systems
MAB51561
102885-912EA 566.89 USD
102885-912
Anti-IL2RA Mouse Monoclonal Antibody [clone: 745520]
Antibodies
Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: 745520
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR