Anti-FAM19A1 Mouse Monoclonal Antibody [clone: 492821]

Supplier: R & D Systems
MAB5154
220021-098EA 577.02 USD
220021-098
Anti-FAM19A1 Mouse Monoclonal Antibody [clone: 492821]
Antibodies
Type: Primary
Antigen Symbol: FAM19A1
Antigen Name: family with sequence similarity 19 (chemokine (C-C motif)-like), member A1
Antigen Synonyms: TAFA-1, TAFA1
Clonality: Monoclonal
Clone: 492821
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR