Anti-ADAMTS15 Mouse Monoclonal Antibody [clone: 561819]

Supplier: R & D Systems
MAB5149
102885-910EA 584.25 USD
102885-910
Anti-ADAMTS15 Mouse Monoclonal Antibody [clone: 561819]
Antibodies
Type: Primary
Antigen: ADAMTS15
Clonality: Monoclonal
Clone: 561819
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR