Anti-FAM19A5 Rat Monoclonal Antibody [clone: 463102]

Supplier: R & D Systems
MAB5148
220021-096EA 594.37 USD
220021-096
Anti-FAM19A5 Rat Monoclonal Antibody [clone: 463102]
Antibodies
Type: Primary
Antigen Symbol: FAM19A5
Antigen Name: family with sequence similarity 19 (chemokine (C-C motif)-like), member A5
Antigen Synonyms: QLLK5208, TAFA-5, TAFA5, UNQ5208
Clonality: Monoclonal
Clone: 463102
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR