Anti-MCPT1 Rat Monoclonal Antibody [clone: 285008]

Supplier: R & D Systems
MAB5146
220021-094EA 594.37 USD
220021-094
Anti-MCPT1 Rat Monoclonal Antibody [clone: 285008]
Antibodies
Type: Primary
Antigen Symbol: MCPT1
Antigen Name: mast cell protease 1
Antigen Synonyms: AV080368, Mcp-1
Clonality: Monoclonal
Clone: 285008
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR