Anti-UMOD Mouse Monoclonal Antibody [clone: 877914]

Supplier: R & D Systems
MAB5144
102885-908EA 594.37 USD
102885-908
Anti-UMOD Mouse Monoclonal Antibody [clone: 877914]
Antibodies
Type: Primary
Antigen: UMOD
Clonality: Monoclonal
Clone: 877914
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR