Anti-SMOC2 Mouse Monoclonal Antibody [clone: 667713]

Supplier: R & D Systems
MAB5140
102885-906EA 600.15 USD
102885-906
Anti-SMOC2 Mouse Monoclonal Antibody [clone: 667713]
Antibodies
Type: Primary
Antigen: SMOC2
Clonality: Monoclonal
Clone: 667713
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR