Anti-VAMP2 Mouse Monoclonal Antibody [clone: 541405]

Supplier: R & D Systems
MAB5136
102885-904EA 594.37 USD
102885-904
Anti-VAMP2 Mouse Monoclonal Antibody [clone: 541405]
Antibodies
Type: Primary
Antigen: VAMP2
Clonality: Monoclonal
Clone: 541405
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR