Anti-PRKCE Mouse Monoclonal Antibody [clone: 666823]

Supplier: R & D Systems
MAB5134
102885-902EA 561.11 USD
102885-902
Anti-PRKCE Mouse Monoclonal Antibody [clone: 666823]
Antibodies
Type: Primary
Antigen: PRKCE
Clonality: Monoclonal
Clone: 666823
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR