Anti-MMP3 Mouse Monoclonal Antibody [clone: 50647]

Supplier: R & D Systems
MAB513
220021-093EA 772.25 USD
220021-093
Anti-MMP3 Mouse Monoclonal Antibody [clone: 50647]
Antibodies
Type: Primary
Antigen: MMP3
Clonality: Monoclonal
Clone: 50647
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR