Anti-CRKL Mouse Monoclonal Antibody [clone: 515405]

Supplier: R & D Systems
MAB5127
220021-092EA 616.06 USD
220021-092
Anti-CRKL Mouse Monoclonal Antibody [clone: 515405]
Antibodies
Type: Primary
Antigen: CRKL
Clonality: Monoclonal
Clone: 515405
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR