Anti-DGKE Mouse Monoclonal Antibody [clone: 670914]

Supplier: R & D Systems
MAB5125
102885-900EA 594.37 USD
102885-900
Anti-DGKE Mouse Monoclonal Antibody [clone: 670914]
Antibodies
Type: Primary
Antigen: DGKE
Clonality: Monoclonal
Clone: 670914
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR