Anti-IRF8 Mouse Monoclonal Antibody [clone: 809926]

Supplier: R & D Systems
MAB5117
102885-898EA 594.37 USD
102885-898
Anti-IRF8 Mouse Monoclonal Antibody [clone: 809926]
Antibodies
Simple Western

Type: Primary
Antigen: IRF8
Clonality: Monoclonal
Clone: 809926
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR