Anti-CHI3L2 Mouse Monoclonal Antibody [clone: 632010]

Supplier: R & D Systems
MAB5116
102885-896EA 594.37 USD
102885-896
Anti-CHI3L2 Mouse Monoclonal Antibody [clone: 632010]
Antibodies
Type: Primary
Antigen: CHI3L2
Clonality: Monoclonal
Clone: 632010
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR