Anti-REG3B Rat Monoclonal Antibody [clone: 518630]

Supplier: R & D Systems
MAB5110
220021-091EA 594.37 USD
220021-091
Anti-REG3B Rat Monoclonal Antibody [clone: 518630]
Antibodies
Type: Primary
Antigen: REG3B
Clonality: Monoclonal
Clone: 518630
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR