Anti-NINJ1 Mouse Monoclonal Antibody [clone: 758943]

Supplier: R & D Systems
MAB5105
102885-892EA 600.15 USD
102885-892
Anti-NINJ1 Mouse Monoclonal Antibody [clone: 758943]
Antibodies
Type: Primary
Antigen Symbol: NINJ1
Antigen Name: ninjurin 1
Antigen Synonyms: NIN1, NINJURIN
Clonality: Monoclonal
Clone: 758943
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR