Anti-FAM19A4 Mouse Monoclonal Antibody [clone: 480103]

Supplier: R & D Systems
MAB5099
220021-088EA 561.11 USD
220021-088
Anti-FAM19A4 Mouse Monoclonal Antibody [clone: 480103]
Antibodies
Type: Primary
Antigen Symbol: FAM19A4
Antigen Name: family with sequence similarity 19 (chemokine (C-C motif)-like), member A4
Antigen Synonyms: TAFA-4, TAFA4
Clonality: Monoclonal
Clone: 480103
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR